siRNA Details
| Gene Symbol: Cemip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759616.1 | XP_008757838.1 | 6 | 1249 | ||
| XM_218855.8 | XP_218855.4 | 6 | 1118 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cemip| Gene Symbol: Cemip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759616.1 | XP_008757838.1 | 6 | 1249 | ||
| XM_218855.8 | XP_218855.4 | 6 | 1118 | |||