siRNA Details
| Gene Symbol: Ccdc90b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024885.1 | NP_001020056.1 | 7 | 870 | ||
| GenBank mRNA | BC097480.1 | - | 7 | 870 | 7458611 | |
| FQ215669.1 | - | 7 | 572 | |||
| FQ218235.1 | - | 7 | 585 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc90b| Gene Symbol: Ccdc90b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024885.1 | NP_001020056.1 | 7 | 870 | ||
| GenBank mRNA | BC097480.1 | - | 7 | 870 | 7458611 | |
| FQ215669.1 | - | 7 | 572 | |||
| FQ218235.1 | - | 7 | 585 | |||