siRNA Details
Gene Symbol: Tenm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191628.1 | NP_001178557.1 | 7 | 1221 | ||
XM_008759718.1 | XP_008757940.1 | 10 | 2936 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tenm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191628.1 | NP_001178557.1 | 7 | 1221 | ||
XM_008759718.1 | XP_008757940.1 | 10 | 2936 |