siRNA Details
| Gene Symbol: Ints4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191629.1 | NP_001178558.1 | 10, 11 | 1226 | ||
| GenBank mRNA | FQ218032.1 | - | 8, 9 | 880 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ints4| Gene Symbol: Ints4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191629.1 | NP_001178558.1 | 10, 11 | 1226 | ||
| GenBank mRNA | FQ218032.1 | - | 8, 9 | 880 | ||