siRNA Details
| Gene Symbol: Prkrir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191630.1 | NP_001178559.1 | 5 | 924 | ||
| XM_008759719.1 | XP_008757941.1 | 7 | 1372 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkrir| Gene Symbol: Prkrir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191630.1 | NP_001178559.1 | 5 | 924 | ||
| XM_008759719.1 | XP_008757941.1 | 7 | 1372 | |||