siRNA Details
| Gene Symbol: Trim21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082572.1 | NP_001076041.1 | 1 | 183 | ||
| XM_006229884.2 | XP_006229946.1 | 2 | 304 | |||
| XM_006229885.2 | XP_006229947.1 | 2 | 304 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim21| Gene Symbol: Trim21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082572.1 | NP_001076041.1 | 1 | 183 | ||
| XM_006229884.2 | XP_006229946.1 | 2 | 304 | |||
| XM_006229885.2 | XP_006229947.1 | 2 | 304 | |||