siRNA Details
| Gene Symbol: Trim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014023.1 | NP_001014045.1 | 3, 4 | 617 | ||
| XM_006229912.2 | XP_006229974.1 | 3, 4 | 1103 | |||
| GenBank mRNA | BC078926.1 | - | 3, 4 | 617 | 7106563 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim5| Gene Symbol: Trim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014023.1 | NP_001014045.1 | 3, 4 | 617 | ||
| XM_006229912.2 | XP_006229974.1 | 3, 4 | 1103 | |||
| GenBank mRNA | BC078926.1 | - | 3, 4 | 617 | 7106563 | |