siRNA Details
| Gene Symbol: Ahi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002277.1 | NP_001002277.1 | 3 | 618 | ||
| GenBank mRNA | AY647140.1 | - | 3 | 618 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ahi1| Gene Symbol: Ahi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002277.1 | NP_001002277.1 | 3 | 618 | ||
| GenBank mRNA | AY647140.1 | - | 3 | 618 | ||