siRNA Details
| Gene Symbol: Ahi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002277.1 | NP_001002277.1 | 19 | 2853 | ||
| GenBank mRNA | AY647140.1 | - | 19 | 2853 | ||
| FQ226565.1 | - | 3134 | ||||
| Gene Symbol: LOC103690149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758641.1 | XP_008756863.1 | 5746 | |||
| XM_008758642.1 | XP_008756864.1 | 5578 | ||||

Ahi1
Genomic Map