siRNA Details
| Gene Symbol: Aldh8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191088.1 | NP_001178017.1 | 4 | 549 | ||
| XM_006227674.2 | XP_006227736.1 | 4 | 584 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh8a1| Gene Symbol: Aldh8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191088.1 | NP_001178017.1 | 4 | 549 | ||
| XM_006227674.2 | XP_006227736.1 | 4 | 584 | |||