siRNA Details
| Gene Symbol: Wee1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012742.1 | NP_001012760.1 | 2 | 762 | ||
| XM_006229974.2 | XP_006230036.1 | 2 | 3492 | |||
| GenBank mRNA | BC090346.1 | - | 2 | 762 | 7306767 | |
| D31838.1 | - | 2 | 961 | |||

Wee1
Genomic Map