siRNA Details
| Gene Symbol: St5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107547.1 | NP_001101017.1 | 6 | 1876 | ||
| XM_006229976.2 | XP_006230038.1 | 7 | 3298 | |||
| XM_006229977.2 | XP_006230039.1 | 7 | 1893 | |||
| XM_006229979.2 | XP_006230041.1 | 6 | 2393 | |||
| XM_008759737.1 | XP_008757959.1 | 5 | 3063 | |||
| XM_008759738.1 | XP_008757960.1 | 7 | 3180 | |||
| XM_008759739.1 | XP_008757961.1 | 8 | 3284 | |||

St5
Genomic Map