siRNA Details
| Gene Symbol: St5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107547.1 | NP_001101017.1 | 19, 20 | 3583 | ||
| XM_006229976.2 | XP_006230038.1 | 20, 21 | 5005 | |||
| XM_006229977.2 | XP_006230039.1 | 20, 21 | 3600 | |||
| XM_006229979.2 | XP_006230041.1 | 19, 20 | 4100 | |||
| XM_008759737.1 | XP_008757959.1 | 18, 19 | 4770 | |||
| XM_008759738.1 | XP_008757960.1 | 20, 21 | 4887 | |||
| XM_008759739.1 | XP_008757961.1 | 21, 22 | 4991 | |||
| XM_008759740.1 | XP_008757962.1 | 17, 18 | 4554 | |||

St5
Genomic Map