siRNA Details
| Gene Symbol: Nfatc2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007692.1 | NP_001007693.1 | 5 | 865 | ||
| GenBank mRNA | BC079050.1 | - | 5 | 865 | 7115323 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfatc2ip| Gene Symbol: Nfatc2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007692.1 | NP_001007693.1 | 5 | 865 | ||
| GenBank mRNA | BC079050.1 | - | 5 | 865 | 7115323 | |