siRNA Details
Gene Symbol: Sez6l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107550.2 | NP_001101020.1 | 9 | 2305 | ||
XM_006230264.1 | XP_006230326.1 | 9 | 2300 | |||
XM_008759881.1 | XP_008758103.1 | 9 | 2211 |
