siRNA Details
| Gene Symbol: Sez6l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107550.2 | NP_001101020.1 | 6 | 1803 | ||
| XM_006230264.1 | XP_006230326.1 | 6 | 1798 | |||
| XM_008759881.1 | XP_008758103.1 | 6 | 1709 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sez6l2| Gene Symbol: Sez6l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107550.2 | NP_001101020.1 | 6 | 1803 | ||
| XM_006230264.1 | XP_006230326.1 | 6 | 1798 | |||
| XM_008759881.1 | XP_008758103.1 | 6 | 1709 | |||