siRNA Details
| Gene Symbol: Itgal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033998.2 | NP_001029170.1 | 30 | 3370 | ||
| XM_006230269.2 | XP_006230331.1 | 30 | 3514 | |||
| GenBank mRNA | BC101849.1 | - | 30 | 3370 | 7938065 | |
| CV076006.1 | - | 2 | 144 | |||

Itgal
Genomic Map