siRNA Details
| Gene Symbol: Itgal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033998.2 | NP_001029170.1 | 28 | 3174 | ||
| XM_006230269.2 | XP_006230331.1 | 28 | 3318 | |||
| GenBank mRNA | BC101849.1 | - | 28 | 3174 | 7938065 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itgal| Gene Symbol: Itgal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033998.2 | NP_001029170.1 | 28 | 3174 | ||
| XM_006230269.2 | XP_006230331.1 | 28 | 3318 | |||
| GenBank mRNA | BC101849.1 | - | 28 | 3174 | 7938065 | |