siRNA Details
| Gene Symbol: LOC102553763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230368.2 | XP_006230430.1 | 2 | 913 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102553763| Gene Symbol: LOC102553763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230368.2 | XP_006230430.1 | 2 | 913 | ||