siRNA Details
| Gene Symbol: Zfp646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759937.1 | XP_008758159.1 | 2 | 1122 | ||
| XM_008759938.1 | XP_008758160.1 | 2 | 865 | |||
| XM_008759939.1 | XP_008758161.1 | 1 | 2460 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp646| Gene Symbol: Zfp646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759937.1 | XP_008758159.1 | 2 | 1122 | ||
| XM_008759938.1 | XP_008758160.1 | 2 | 865 | |||
| XM_008759939.1 | XP_008758161.1 | 1 | 2460 | |||