siRNA Details
| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134859.1 | NP_001128331.1 | 10 | 1964 | ||
| XM_006230387.1 | XP_006230449.1 | 10 | 1898 | |||
| XM_006230388.1 | XP_006230450.1 | 9 | 1465 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec23ip| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134859.1 | NP_001128331.1 | 10 | 1964 | ||
| XM_006230387.1 | XP_006230449.1 | 10 | 1898 | |||
| XM_006230388.1 | XP_006230450.1 | 9 | 1465 | |||