siRNA Details
| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134859.1 | NP_001128331.1 | 17 | 2992 | ||
| XM_006230387.1 | XP_006230449.1 | 17 | 2923 | |||
| XM_006230388.1 | XP_006230450.1 | 16 | 2493 | |||
| GenBank mRNA | BC098824.1 | - | 4 | 817 | ||

Sec23ip
Genomic Map