siRNA Details
| Gene Symbol: RGD1305014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014025.1 | NP_001014047.1 | 5 | 912 | ||
| XM_008759885.1 | XP_008758107.1 | 5 | 2114 | |||
| GenBank mRNA | BC083846.1 | - | 5 | 912 | 7121351 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1305014| Gene Symbol: RGD1305014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014025.1 | NP_001014047.1 | 5 | 912 | ||
| XM_008759885.1 | XP_008758107.1 | 5 | 2114 | |||
| GenBank mRNA | BC083846.1 | - | 5 | 912 | 7121351 | |