siRNA Details
Gene Symbol: RGD1305014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014025.1 | NP_001014047.1 | 4 | 622 | ||
XM_008759885.1 | XP_008758107.1 | 4 | 1824 | |||
GenBank mRNA | BC083846.1 | - | 4 | 622 | 7121351 |
