siRNA Details
Gene Symbol: Ikzf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107555.1 | NP_001101025.1 | 5 | 647 | ||
XM_006230401.1 | XP_006230463.1 | 4 | 484 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ikzf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107555.1 | NP_001101025.1 | 5 | 647 | ||
XM_006230401.1 | XP_006230463.1 | 4 | 484 |