siRNA Details
| Gene Symbol: Dcun1d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024886.1 | NP_001020057.1 | 3 | 911 | ||
| XM_008759750.1 | XP_008757972.1 | 3 | 837 | |||
| GenBank mRNA | BC097462.1 | - | 3 | 911 | 7457575 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcun1d3| Gene Symbol: Dcun1d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024886.1 | NP_001020057.1 | 3 | 911 | ||
| XM_008759750.1 | XP_008757972.1 | 3 | 837 | |||
| GenBank mRNA | BC097462.1 | - | 3 | 911 | 7457575 | |