siRNA Details
| Gene Symbol: Edrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107557.1 | NP_001101027.1 | 21, 22 | 3313 | ||
| XM_006230436.2 | XP_006230498.1 | 22, 23 | 3961 | |||
| XM_008759889.1 | XP_008758111.1 | 22, 23 | 3963 | |||

Edrf1
Genomic Map