siRNA Details
| Gene Symbol: Uros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012068.1 | NP_001012068.1 | 8 | 698 | ||
| XM_008759890.1 | XP_008758112.1 | 7 | 899 | |||
| XM_008759891.1 | XP_008758113.1 | 7 | 594 | |||
| XM_008759892.1 | XP_008758114.1 | 7 | 867 | |||
| GenBank mRNA | BC083730.1 | - | 8 | 698 | 7191339 | |

Uros
Genomic Map