siRNA Details
| Gene Symbol: Fank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008347.1 | NP_001008348.1 | 5 | 519 | ||
| XM_006230439.2 | XP_006230501.1 | 5 | 564 | |||
| GenBank mRNA | BC082095.1 | - | 5 | 519 | 7134234 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fank1| Gene Symbol: Fank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008347.1 | NP_001008348.1 | 5 | 519 | ||
| XM_006230439.2 | XP_006230501.1 | 5 | 564 | |||
| GenBank mRNA | BC082095.1 | - | 5 | 519 | 7134234 | |