siRNA Details
| Gene Symbol: Dock1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143858.1 | NP_001137330.1 | 2 | 197 | ||
| XM_008759987.1 | XP_008758209.1 | 2 | 190 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dock1| Gene Symbol: Dock1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143858.1 | NP_001137330.1 | 2 | 197 | ||
| XM_008759987.1 | XP_008758209.1 | 2 | 190 | |||