siRNA Details
| Gene Symbol: Gpr123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760017.1 | XP_008758239.1 | 3, 4 | 1713 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr123| Gene Symbol: Gpr123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760017.1 | XP_008758239.1 | 3, 4 | 1713 | ||
Set Membership: |
|