siRNA Details
| Gene Symbol: Tubgcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107560.2 | NP_001101030.1 | 16 | 2547 | ||
| XM_006230457.1 | XP_006230519.1 | 16 | 2557 | |||
| XM_006230458.1 | XP_006230520.1 | 17 | 2734 | |||
| XM_006230459.2 | XP_006230521.1 | 18 | 2868 | |||
| XM_006230460.1 | XP_006230522.1 | 17 | 2720 | |||
| XM_006230461.1 | XP_006230523.1 | 13 | 1970 | |||
| GenBank mRNA | BC166857.1 | - | 16 | 2547 | 9100036 | |

Tubgcp2
Genomic Map