siRNA Details
| Gene Symbol: Fcho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191632.1 | NP_001178561.1 | 5 | 559 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fcho2| Gene Symbol: Fcho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191632.1 | NP_001178561.1 | 5 | 559 | ||