siRNA Details
| Gene Symbol: Ano1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107564.1 | NP_001101034.1 | 11 | 1200 | ||
| XM_006230780.2 | XP_006230842.1 | 10 | 1202 | |||
| XM_008760118.1 | XP_008758340.1 | 10 | 1203 | |||
| XM_008760119.1 | XP_008758341.1 | 10 | 1120 | |||

Ano1
Genomic Map