siRNA Details
Gene Symbol: Tpcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107566.1 | NP_001101036.1 | 3, 4 | 283 | ||
XM_006230784.2 | XP_006230846.1 | 3, 4 | 306 | |||
GenBank mRNA | BC166782.1 | - | 3, 4 | 219 | 7935012 |
