siRNA Details
Gene Symbol: Tpcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107566.1 | NP_001101036.1 | 17, 18 | 1650 | ||
XM_006230784.2 | XP_006230846.1 | 18, 19 | 1739 | |||
GenBank mRNA | BC166782.1 | - | 18, 19 | 1714 | 7935012 |
