siRNA Details
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107569.1 | NP_001101039.1 | 15, 16 | 1648 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107569.1 | NP_001101039.1 | 15, 16 | 1648 |