siRNA Details
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107569.1 | NP_001101039.1 | 7 | 600 | ||
XM_008760137.1 | XP_008758359.1 | 7 | 618 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107569.1 | NP_001101039.1 | 7 | 600 | ||
XM_008760137.1 | XP_008758359.1 | 7 | 618 |