siRNA Details
| Gene Symbol: Rela | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199267.2 | NP_954888.1 | 3 | 216 | ||
| GenBank mRNA | AF079314.1 | - | 3 | 230 | ||
| AY307375.1 | - | 2 | 155 | |||
| BC079457.1 | - | 3 | 216 | 7121610 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rela| Gene Symbol: Rela | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199267.2 | NP_954888.1 | 3 | 216 | ||
| GenBank mRNA | AF079314.1 | - | 3 | 230 | ||
| AY307375.1 | - | 2 | 155 | |||
| BC079457.1 | - | 3 | 216 | 7121610 | ||