siRNA Details
| Gene Symbol: Pcnxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230947.2 | XP_006231009.1 | 23 | 4311 | ||
| XM_006230948.2 | XP_006231010.1 | 23 | 4308 | |||
| XM_006230950.2 | XP_006231012.1 | 23 | 4281 | |||
| XM_006230952.2 | XP_006231014.1 | 23 | 4242 | |||
| XM_219515.8 | XP_219515.4 | 23 | 4272 | |||

Pcnxl3
Genomic Map