siRNA Details
| Gene Symbol: Map3k11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013150.1 | NP_001013168.1 | 9 | 2588 | ||
| GenBank mRNA | BC081952.1 | - | 9 | 2588 | 7122664 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k11| Gene Symbol: Map3k11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013150.1 | NP_001013168.1 | 9 | 2588 | ||
| GenBank mRNA | BC081952.1 | - | 9 | 2588 | 7122664 | |