siRNA Details
| Gene Symbol: Mrpl49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047883.1 | NP_001041348.1 | 1 | 53 | ||
| GenBank mRNA | AY325168.1 | - | 1 | 53 | ||
| FQ209431.1 | - | 18 | ||||
| FQ212546.1 | - | 32 | ||||
| FQ235268.1 | - | 32 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl49| Gene Symbol: Mrpl49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047883.1 | NP_001041348.1 | 1 | 53 | ||
| GenBank mRNA | AY325168.1 | - | 1 | 53 | ||
| FQ209431.1 | - | 18 | ||||
| FQ212546.1 | - | 32 | ||||
| FQ235268.1 | - | 32 | ||||