siRNA Details
| Gene Symbol: Fermt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127543.1 | NP_001121015.1 | 11 | 1398 | ||
| XM_006230811.2 | XP_006230873.1 | 11 | 1889 | |||
| GenBank mRNA | BC166607.1 | - | 11 | 1398 | 5598287 | |
| FQ210530.1 | - | 2 | 162 | |||

Fermt3
Genomic Map