siRNA Details
| Gene Symbol: Fermt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127543.1 | NP_001121015.1 | 3 | 288 | ||
| XM_006230811.2 | XP_006230873.1 | 3 | 779 | |||
| GenBank mRNA | BC166607.1 | - | 3 | 288 | 5598287 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fermt3| Gene Symbol: Fermt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127543.1 | NP_001121015.1 | 3 | 288 | ||
| XM_006230811.2 | XP_006230873.1 | 3 | 779 | |||
| GenBank mRNA | BC166607.1 | - | 3 | 288 | 5598287 | |