siRNA Details
Gene Symbol: Ints5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107576.1 | NP_001101046.1 | 2 | 1307 | ||
GenBank mRNA | FQ226934.1 | - | 2 | 1297 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ints5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107576.1 | NP_001101046.1 | 2 | 1307 | ||
GenBank mRNA | FQ226934.1 | - | 2 | 1297 |