siRNA Details
| Gene Symbol: Ms4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107578.1 | NP_001101048.1 | 5 | 645 | ||
| XM_006231089.2 | XP_006231151.1 | 6 | 817 | |||
| GenBank mRNA | BC098745.1 | - | 5 | 1763 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a1| Gene Symbol: Ms4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107578.1 | NP_001101048.1 | 5 | 645 | ||
| XM_006231089.2 | XP_006231151.1 | 6 | 817 | |||
| GenBank mRNA | BC098745.1 | - | 5 | 1763 | ||