siRNA Details
| Gene Symbol: Olr337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000507.1 | NP_001000507.1 | 1 | 740 | ||
| GenBank mRNA | AF091562.1 | - | 1 | 548 | ||
| AF091564.1 | - | 1 | 548 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr337| Gene Symbol: Olr337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000507.1 | NP_001000507.1 | 1 | 740 | ||
| GenBank mRNA | AF091562.1 | - | 1 | 548 | ||
| AF091564.1 | - | 1 | 548 | |||