siRNA Details
| Gene Symbol: Gna14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013151.1 | NP_001013169.1 | 4 | 1007 | ||
| XM_008760288.1 | XP_008758510.1 | 5 | 1644 | |||
| GenBank mRNA | BC090316.1 | - | 4 | 1007 | 7305112 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gna14| Gene Symbol: Gna14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013151.1 | NP_001013169.1 | 4 | 1007 | ||
| XM_008760288.1 | XP_008758510.1 | 5 | 1644 | |||
| GenBank mRNA | BC090316.1 | - | 4 | 1007 | 7305112 | |