siRNA Details
| Gene Symbol: Pdcd1lg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107582.2 | NP_001101052.2 | 5 | 900 | ||
| XM_006231209.2 | XP_006231271.1 | 4 | 952 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdcd1lg2| Gene Symbol: Pdcd1lg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107582.2 | NP_001101052.2 | 5 | 900 | ||
| XM_006231209.2 | XP_006231271.1 | 4 | 952 | |||