siRNA Details
| Gene Symbol: RGD1311595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231257.2 | XP_006231319.1 | 7 | 4994 | ||
| XM_219779.8 | XP_219779.4 | 8 | 5084 | |||
| GenBank mRNA | BC105763.1 | - | 1 | 163 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311595| Gene Symbol: RGD1311595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231257.2 | XP_006231319.1 | 7 | 4994 | ||
| XM_219779.8 | XP_219779.4 | 8 | 5084 | |||
| GenBank mRNA | BC105763.1 | - | 1 | 163 | ||